missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUBG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596533
This item is not returnable.
View return policy
Description
Immunogen sequence: TSISSLFERT CRQYDKLRKR EAFLEQFRKE DMFKDNFDEM DTSREIVQQL IDEYHAATRP DYISWGTQEQ Positive Samples: Mouse testis, Rat heart; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center.
This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7.
Specifications
| TUBG1 | |
| Polyclonal | |
| Unconjugated | |
| TUBG1 | |
| 1500010O08Rik; AI451582; AI503389; CDCBM4; gamma-1-tubulin; Gamma-tubulin complex component 1; GCP-1; Tubg; TUBG1; TUBGCP1; tubulin gamma 1; tubulin gamma-1 chain; tubulin, gamma 1; tubulin, gamma polypeptide | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 103733, 252921, 7283 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.93 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23258, P83887, P83888 | |
| TUBG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 382-451 of human ?-Tubulin (NP_0010612). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction