Learn More
Thermo Scientific™ Tubes and Ultra Clear Caps, strips of 8, Sustain™ Series
GREENER_CHOICE
Product Code. 30179702 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
120 Strips
Unit Size:
Pack of 120
Product Code. Quantity unitSize
30179702 120 Strips Pack of 120
1 options
Product Code. 30179702 Supplier Thermo Scientific™ Supplier No. AB1183SS
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Thermo Scientific Tubes and Ultra Clear Caps, strips of 8, Sustain Series, are designed for PCR and qPCR applications and are suitable for 0.2 mL thermal cycler blocks and 96-well V-bottom thermal cyclers.
Thermo Scientific Tubes and Ultra Clear Caps, strips of 8, Sustain Series, are designed for PCR and qPCR applications and are suitable for 0.2 mL thermal cycler blocks and 96-well V-bottom thermal cyclers. The 8-strip tubes can be divided into individual tubes.
Features and benefits
  • Suitable for 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Certified DNase-, RNase-, human DNA-, and PCR inhibitor-free
A breakthrough in sustainable PCR technology
Thermo Scientific Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
  • Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
  • Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
  • Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific PCR plastics
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free
Color Natural
Quantity 120 Strips
Material Polypropylene
Closure Type Optical Strip Cap
Autoclavable Autoclavable
Closure Included Yes
Green Features ISCC PLUS