missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- PTFE construction for chemical resistance
- Secure, screw-type connection
- Suitable for FlexTube tubing
Specifications
Specifications
| Material | PTFE |
| For Use With | FlexTube GL25 Tubing |
| Packaging Quantity | 2 Pk. |
| Color | Black |
| Product Type | Screw Connection |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?