missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubercidin, MedChemExpress
Description
Tubercidin inhibits the growth of Streptococcus faecalis (8043) with an IC50 of 0.02 μM. Tubercidin inhibits polymerases by incorporating DNA or RNA, thereby inhibiting DNA replication, RNA and protein synthesis. Tubercidin is a weak inhibitor of adenosine phosphorylase, and interferes with the phosphorylation of adenosine and AMP. Tubercidin has antiviral activity.
Specifications
Specifications
| Chemical Name or Material | Tubercidin |
| Physical Form | Solid |
| CAS | 69-33-0 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C11H14N4O4 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | 7-Deazaadenosine |
| Solubility Information | DMSO : ≥ 30 mg/mL (112.68 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?