missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubeimoside II, MedChemExpress
Description
Tubeimoside II(Tubeimoside-B) is a natural analogue of oleanane type of triterpenoid saponin
Specifications
Specifications
| Chemical Name or Material | Tubeimoside II |
| CAS | 115810-12-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C63H98O30 |
| Quantity | 10 mg |
| Synonym | Tubeimoside-B |
| Solubility Information | DMSO : 100 mg/mL (74.88 mM; Need ultrasonic) |
| SMILES | OC1C(C(OC(OCC(O)C2O)C2OC(OC(C)C(OC(CC(O)(CC(O3)=O)C)=O)C4OC(OCC(O)C5O)C5O)C4O)=O)(CCC(C)(C)C6)C6C7=CCC8C(C)(CCC9C8(CC(O)C(OC(OC(CO)C(O)C%10O)C%10OC(OCC3C%11O)C%11O)C9(C)CO)C)C7(C)C1 |
| Molecular Weight (g/mol) | 1335.43 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?