missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Tubeimoside I

Product Code. 16444907
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444907 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444907 Supplier MedChemExpress Supplier No. HYN0890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tubeimoside I(Lobatoside-H) is an extract from Chinese herbal medicine Bolbostemma paniculatum (MAXIM.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 1319.44 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 102040-03-9
Product Type Natural Products
Purity 99.07%
Solubility DMSO : 100 mg/mL (75.79 mM∣Need ultrasonic)
Synonym Tubeimoside-1∣ Lobatoside-H
Target Apoptosis
Quantity 50 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.