Learn More
TUBE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544635
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: HLHHYLQVEG MEESCFTEAV SSLSALIQEY DQLDATKNMP VQDLPRLSIA Sequence homology: Dog: 85%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
TUBE1 Polyclonal antibody specifically detects TUBE1 in Drosophila, Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
TUBE1 | |
Polyclonal | |
Unconjugated | |
TUBE1 | |
2310061K05Rik; AI551343; dJ142L7.2; epsilon-tubulin; epsilon-tubulin 1; FLJ22589; FLJ44203; RP1-142L7.1; TUBE; Tube1; Tubulin; tubulin epsilon 1; tubulin epsilon chain; tubulin, epsilon 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UJT0 | |
TUBE1 | |
Synthetic peptide directed towards the middle region of human TUBE1 | |
100 μL | |
Primary | |
Drosophila, Human | |
Antibody | |
IgG |