missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBE1 Monoclonal Antibody (TUB-11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120249
This item is not returnable.
View return policy
Description
MA1-20249 has been successfully used in ELISA and Western blot procedures. The MA1-20249 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues G(352) N V Q I S D L R R N I E R L(366) of human epsilon tubulin.
Microtubules constitute one of the major components of eukaryotic cells cytoskeleton and are involved in many essential processes, including cell division, ciliary and flagellar motility and intracellular transport. Microtubules of the eukaryotic cytoskeleton are composed of a heterodimer of α- and β-tubulin. In addition to α- and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven. Most of these tubulins have distinct subcellular localization and an emerging, diverse set of functions.
Specifications
| TUBE1 | |
| Monoclonal | |
| 2.4 mg/mL | |
| TUBE1 | |
| RP1-142L7.1, FLJ22589, FLJ44203, TUBE, dJ142L7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 51175, 71924 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| TUB-11 | |
| Unconjugated | |
| Q9UJT0, Q9D6T1 | |
| TUBE1 | |
| Synthetic peptide, conjugated to KLH, corresponding to residues G(352) N V Q I S D L R R N I E R L(366) of human epsilon tubulin. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction