Learn More
Scientific Industries SI™ Tube Rack
Accommodates up to 10 tubes of any material. up to 29mm diameter. Scientific Industries SI™ Tube Rack is for use with Vortex-Genie™ MAX.
Brand:  Scientific Industries SI™ SI-MX020
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15547355
  • £266.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • Holds up to 10 tubes
  • For use with Vortex-Genie™ MAX
Specifications
Specifications
29 mm dia. Tubes
20
2/Pk.