missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ AutoLys 24-tube Rack

Product Code. 15117386
Click to view available options
Positions:
24
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 15117386

Brand: Hamilton™ 235459

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SBS-format, 24-well rack designed for easy lysis processing. Hamilton™ AutoLys 24-tube Rack is a highly complementary accessory component for Hamilton™ AutoLys tubes.

  • Ideal for automated barcode reading from the base.
  • Special rack design holds tubes in position during lift and lock step.

Specifications

Product Type AutoLys Tube Rack
Color Clear
Positions 24
For Use With (Equipment) Hamilton™ AutoLys tubes
No. per Case 25
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.