Learn More
Cytiva Tube Rack
Product Code. 12465710 Shop All Cytiva Products
Click to view available options
Holds:
Tubes (50 mL)
Unit Size:
Each
Product Code. 12465710
Brand: Cytiva undefined
Only null left
Add to basket
Add to basket
Holds up to 55 tubes
Holds up to 55 tubes. For use with Fraction collector F9-C and ÅKTA avant.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube Rack
Holds Tubes (50 mL)
Positions 55
Quantity 1 Each