Learn More
Cytiva Tube Holder
Shop All Cytiva Products
Click to view available options
For Use With:
95 Tubes
Frac-900
Unit Size:
1 set
Pack of 2
Description
The tube guide can be placed at different heights to accommodate different length of tubes.
Specifications
Specifications
| Type | Tube Holder |
| Packaging Quantity | 2 Pack |
| Includes | Tube Holder and Guide |
| For Use With | Frac-900 |