missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Tube Cleaning Brush

The Tube Cleaning Brush is ideal for cleaning zone samplers.

Brand:  Buerkle™ 5304-0040

48.42 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.



Code : A0

Additional Details : Weight : 0.09300kg

 View more versions of this product

Product Code. 10108161

  • £58.85 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Manufactured from PVC and stainless steel
  • Available in a range of lengths
Specifications

Specifications

Cleaning Brush
Used for Cleaning
40 cm
25 mm
Zone Sampler
PVC, Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers