missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Tube Cap, Straight Plugs

Product Code. 12374039
Change view
Click to view available options
Quantity:
1000/Pk.
Unit Size:
Pack of 20000
Product Code. Quantity unitSize
12374039 1000/Pk. Pack of 20000
1 options
This item is not returnable. View return policy

Product Code. 12374039

Brand: Gosselin™ B9003

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Corning™ Gosselin™ Tube Cap, Straight Plugs are produced via injection molding using virgin plastics that comply with current environmental standards and are vertically ribbed for easier grip.

  • All closures and tubes are produced with high-tech equipment using injection molding technology.
  • The raw plastics used are virgin and comply with the current environmental standards.
  • Each production lot is subject to inspection throughout the entire production process according to the standard references, which are reinforced by Gosselin’s own specifications.
  • All closures are vertically ribbed for easier grip.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Straight Plug
Material LDPE
Color White
No. per Pack 1000/Pk.
No. per Case 20,000/Cs.
Quantity 1000/Pk.
For Use With (Equipment) Tube code: TL40
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.