missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBAL3 Antibody (OTI4C5), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74696MFV610
This item is not returnable.
View return policy
Description
TUBAL3 Monoclonal antibody specifically detects TUBAL3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry
Specifications
| TUBAL3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 79861 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| OTI4C5 | |
| Flow Cytometry | |
| FLJ21665, MGC119347, MGC119349, tubulin alpha chain-like 3, tubulin, alpha-like 3 | |
| Human recombinant protein fragment corresponding to amino acids 150-446 of human TUBAL3(NP_079079) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction