missing translation for 'onlineSavingsMsg'
Learn More

Tubacin, MedChemExpress

Product Code. 16438377
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16438377

Brand: Medchem Express HY13428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tubacin is a potent and selective inhibitor of HDAC6, with an IC50 value of 4 nM and approximately 350-fold selectivity over HDAC1. Tubacin also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Tubacin is a potent and selective inhibitor of HDAC6, with an IC50 value of 4 nM and approximately 350-fold selectivity over HDAC1. Tubacin also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Specifications

Chemical Name or Material Tubacin
CAS 537049-40-4
Color White
Purity Grade Notes Research
Molecular Formula C41H43N3O7S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (138.53 mM; Need ultrasonic)
SMILES OCC(C=C1)=CC=C1[C@@H]2C[C@H](CSC3=NC(C4=CC=CC=C4)=C(C5=CC=CC=C5)O3)O[C@H](C6=CC=C(NC(CCCCCCC(NO)=O)=O)C=C6)O2
Molecular Weight (g/mol) 721.86
Formula Weight 721.86
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.