missing translation for 'onlineSavingsMsg'
Learn More

Tubacin, MedChemExpress

Product Code. 16460546
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460546 20 mg 20mg
16438377 5 mg 5mg
16415688 10 mg 10mg
16401349 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16460546 Supplier Medchem Express Supplier No. HY13428
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Tubacin is a potent and selective inhibitor of HDAC6, with an IC50 value of 4 nM and approximately 350-fold selectivity over HDAC1. Tubacin also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Tubacin is a potent and selective inhibitor of HDAC6, with an IC50 value of 4 nM and approximately 350-fold selectivity over HDAC1. Tubacin also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Specifications

Chemical Name or Material Tubacin
CAS 537049-40-4
Color White
Purity Grade Notes Research
Molecular Formula C41H43N3O7S
Quantity 20 mg
Solubility Information DMSO : 100 mg/mL (138.53 mM; Need ultrasonic)
SMILES OCC(C=C1)=CC=C1[C@@H]2C[C@H](CSC3=NC(C4=CC=CC=C4)=C(C5=CC=CC=C5)O3)O[C@H](C6=CC=C(NC(CCCCCCC(NO)=O)=O)C=C6)O2
Molecular Weight (g/mol) 721.86
Formula Weight 721.86
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.