Learn More
Invitrogen™ TUBA8 Monoclonal Antibody (2D6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517193
Description
MA5-17193 targets TUBA8 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Rat samples. The MA5-17193 immunogen is purified recombinant fragment of human TUBA8 expressed in E. Coli. MA5-17193 detects TUBA8 which has a predicted molecular weight of approximately 50kDa.
This gene encodes a member of the alpha tubulin protein family. Alpha tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene are associated with polymicrogyria and optic nerve hypoplasia. Alternate splicing results in multiple transcript variants.
Specifications
| TUBA8 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q6AY56, Q9JJZ2, Q9NY65 | |
| TUBA8 | |
| Purified recombinant fragment of human TUBA8 expressed in E. Coli. | |
| RUO | |
| 500377, 51807, 53857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2D6 | |
| Unconjugated | |
| TUBA8 | |
| alpha tubulin 8; alpha-tubulin 8; TUBA8; TUBAL2; tubulin alpha 8; tubulin alpha chain-like 2; Tubulin alpha-8 chain; tubulin, alpha 8 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.