missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17928-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the Tubby family of bipartite transcription factors. The encoded protein may play a role in obesity and sensorineural degradation. The crystal structure has been determined for a similar protein in mouse, and it functions as a membrane-bound transcription regulator that translocates to the nucleus in response to phosphoinositide hydrolysis. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| TUB | |
| Polyclonal | |
| Unconjugated | |
| TUB | |
| rd5, TUB, tubby homolog (mouse), Tubby protein homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22141, 25609, 7275 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88808, P50586, P50607 | |
| TUB | |
| TUB Fusion Protein Ag12354 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction