missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTYH2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41177B
This item is not returnable.
View return policy
Description
TTYH2 Polyclonal antibody specifically detects TTYH2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TTYH2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| C17orf29, hTTY2, Protein Tweety Homolog 2, Tweety (Drosophila) Homolog 2, Tweety Family Member 2, Tweety Homolog 2, Tweety Homolog 2 (Drosophila) | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human TTYH2. The immunogen is located within amino acids 60 - 110 of TTYH2. Amino Acid Squence: RRDDAVQTKQHHS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 94015 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction