missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTP Antibody (OTI8B5), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74681PECY55
This item is not returnable.
View return policy
Description
TTP Monoclonal specifically detects TTP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| TTP | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| ZFP36 | |
| Full length recombinant protein of human TTP (NP_003398) produced in HEK293T cells. | |
| 0.1 mL | |
| 7538.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI8B5 | |
| Flow Cytometry | |
| EZNFTranscription factor 17, HKL1Zinc finger protein eZNF, KID-1, KID1TCF-17, TCF17, zinc finger protein 354A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction