missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTP Antibody (OTI8B5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74681CL1
This item is not returnable.
View return policy
Description
TTP Monoclonal antibody specifically detects TTP in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| TTP | |
| Monoclonal | |
| CoraFluor 1 | |
| EZNFTranscription factor 17, HKL1Zinc finger protein eZNF, KID-1, KID1TCF-17, TCF17, zinc finger protein 354A | |
| Full length human recombinant protein of human ZFP36 (NP_003398) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI8B5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 7538 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction