missing translation for 'onlineSavingsMsg'
Learn More

TTNPB, MedChemExpress

Product Code. p-200101068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499287

Brand: Medchem Express HY15682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively.

TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively.

Specifications

Chemical Name or Material TTNPB
CAS 71441-28-6
Color White
Purity Grade Notes Research
Molecular Formula C24H28O2
Quantity 5 mg
Synonym Ro 13-7410 Arotinoid acid AGN191183
Solubility Information DMSO : 7.58 mg/mL (21.75 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(/C=C(C2=CC=C3C(C)(C)CCC(C)(C)C3=C2)\C)C=C1
Molecular Weight (g/mol) 348.48
Formula Weight 348.48
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.