missing translation for 'onlineSavingsMsg'
Learn More

TTNPB, MedChemExpress

Product Code. 16499287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499287 5 mg 5mg
16476738 10 mg 10mg
16472817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499287 Supplier Medchem Express Supplier No. HY15682
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively.

TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively.

Specifications

Chemical Name or Material TTNPB
CAS 71441-28-6
Color White
Purity Grade Notes Research
Molecular Formula C24H28O2
Quantity 5 mg
Synonym Ro 13-7410 Arotinoid acid AGN191183
Solubility Information DMSO : 7.58 mg/mL (21.75 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(/C=C(C2=CC=C3C(C)(C)CCC(C)(C)C3=C2)\C)C=C1
Molecular Weight (g/mol) 348.48
Formula Weight 348.48
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.