missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTLL6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100050
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TTLL6.
Predicted to enable tubulin binding activity and tubulin-glutamic acid ligase activity. Predicted to be involved in microtubule cytoskeleton organization and protein polyglutamylation. Predicted to act upstream of or within microtubule bundle formation; microtubule severing; and positive regulation of cilium movement. Located in ciliary basal body.
Specifications
| TTLL6 | |
| Polyclonal | |
| Unconjugated | |
| TTLL6 | |
| 4932418K24Rik; D11Moh43e; D11Moh44e; polyglutamylase; RGD1311922; t8130b59; TTL.6; TTLL6; Ttll6-ps1; Tubulin polyglutamylase TTLL6; tubulin tyrosine ligase like 6; tubulin tyrosine ligase-like family member 6; tubulin tyrosine ligase-like family, member 6; tubulin tyrosine ligase-like family, member 6, pseudogene 1; tubulin--tyrosine ligase-like protein 6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 237930, 284076, 287646 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A4Q9E8, Q8N841 | |
| TTLL6 | |
| A synthesized peptide derived from human TTLL6(Accession Q8N841), corresponding to amino acid residues W91-P141. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction