missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTLL12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576352
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
TTLL12 (tubulin tyrosine ligase-like family, member 12) is a 644 amino acid protein that contains one TTL domain. Of the fourteen members of the TTLL family that modify tubulin, TTLL12 is the least characterized member. TTLL12 is highly expressed in a multitude of metastatic prostate cancer cell lines, therefore, it is considered a target for tumor therapy. Overexpression of TTLL12 is suggested to alter chromosome ploidy, whereas downregulation of TTLL12 influence several post-translational modifications of tubulin.
Specifications
| TTLL12 | |
| Polyclonal | |
| Unconjugated | |
| TTLL12 | |
| BC055368; D430005B17; dJ526I14.2; Inactive tubulin--tyrosine ligase-like protein 12; KIAA0153; RGD1305319; TTL domain protein; ttll12; tubulin tyrosine ligase like 12; tubulin tyrosine ligase-like family member 12; tubulin tyrosine ligase-like family, member 12; tubulin--tyrosine ligase-like protein 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 223723, 23170, 300105 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14166, Q3UDE2 | |
| TTLL12 | |
| Recombinant protein of human TTLL12. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction