missing translation for 'onlineSavingsMsg'
Learn More

TTLL1 Antibody (2C6), Novus Biologicals™

Product Code. 18381609 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381609 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381609 Supplier Novus Biologicals Supplier No. H00025809M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TTLL1 Monoclonal antibody specifically detects TTLL1 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2C6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14968
Gene Alias C22orf7, catalytic subunit of neural tubulin polyglutamylase, EC 6.-, HS323M22B, KIAA0173, PGs3, probable tubulin polyglutamylase TTLL1, Tubulin polyglutamylase complex subunit 3, tubulin tyrosine ligase-like 1, tubulin tyrosine ligase-like family, member 1, tubulin-tyrosine ligase, Tubulin--tyrosine ligase-like protein 1
Host Species Mouse
Immunogen TTLL1 (AAH14968, 1 a.a. ~ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAGKVKWVTDIEKSVLINNFEKRGWVQVTENEDWNFYWMSVQTIRNVFSVEAGYRLSDDQIVNHFPNHYELTRKDLMVKNIKRYRKELEKEGSPLAEKDENGKYLYLDFVPVTYMLPADYNLFVEEFRKSPSSTWIMKPCGKAQGKGIFLINKLSQIKKWSRDSKTSSFVSQSNKEAYVISLYINNPLLIGGRKFDLRLYVLVSTYRPLRCYMYKLGFCRFCTVKYTPSTSELDNMFVHPTNVAIQKHGEDYNHIHGGKWTVSNLRLYLESTRGKEVTSKLFDEIHWIIVQSLKAVAPVMNNDKHCFECYGYDIIIDDKLKPWLIEVNASPSLTSSTANDRILKYNLINDTLNIAVPNGEIPDCKWNKSPPKEVLGNYEILYDEELAQGDGADRELRSRQGQSLGPRAGRSRDSGRAVLTTWK
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25809
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.