missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TTL Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566871

Product Code. 15939554

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LQIKHIIRNCL LSVEPAISTK HLPYQSFQLF GFDFMVDEEL KVWLIEVNGA PACAQKLYAE LCQGIVDIAI SSVFPPPDVE QPQTQP Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TTL
Polyclonal
Unconjugated
TTL
2410003M22Rik; 2700049H19Rik; AI848570; Ttl; tubulin tyrosine ligase; tubulin-tyrosine ligase; tubulin--tyrosine ligase
Rabbit
Antigen Affinity Chromatography
RUO
150465
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NG68
TTL
Recombinant Human TTL. Recombinant protein control fragment (Product #RP-107522).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.