missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TTK Polyclonal specifically detects TTK in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | TTK |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CT96, dual specificity protein kinase TTK, EC 2.7.12.1, ESK, FLJ38280, monopolar spindle 1-like 1, MPS1, MPS1L1PYTcancer/testis antigen 96, phosphotyrosine picked threonine kinase, Phosphotyrosine picked threonine-protein kinase, TTK protein kinase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human TTK. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?