missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TTF2 Polyclonal Antibody

Product Code. 16367744
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16367744 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16367744 Supplier Invitrogen™ Supplier No. PA580642

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TTF2.

This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTF2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TTF2
Gene Accession No. Q9UNY4
Gene Alias 4632434F22Rik; 8030447N19; AV218430; F2; HuF2; human factor 2; lodestar homolog; lodestar protein; RNA polymerase II termination factor; Transcription release factor 2; transcription termination factor 2; transcription termination factor, RNA polymerase II; Ttf2; ZGRF6; zinc finger, GRF-type containing 6
Gene Symbols TTF2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Human TTF2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8458
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.