missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | TTF2 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1-2 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672676
|
Novus Biologicals
NBP2-98952-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666225
|
Novus Biologicals
NBP2-98952-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTF2 Polyclonal antibody specifically detects TTF2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| TTF2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 3.6.1, EC 3.6.4.-, F2, HuF2lodestar protein, human factor 2, Lodestar homolog, RNA polymerase II termination factor, Transcription release factor 2, transcription termination factor 2, transcription termination factor, RNA polymerase II | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TTF2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000, Immunoprecipitation 1-2 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| 8458 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title