missing translation for 'onlineSavingsMsg'
Learn More

TTF2 Antibody (2B6), Novus Biologicals™

Product Code. 18340239 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340239 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340239 Supplier Novus Biologicals Supplier No. H00008458M12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TTF2 Monoclonal antibody specifically detects TTF2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTF2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2B6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003585
Gene Alias EC 3.6.1, EC 3.6.4.-, F2, HuF2lodestar protein, human factor 2, Lodestar homolog, RNA polymerase II termination factor, Transcription release factor 2, transcription termination factor 2, transcription termination factor, RNA polymerase II
Host Species Mouse
Immunogen TTF2 (NP_003585, 385 a.a. ~ 509 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QFPDRSVQRKVSPASGVSKKVEPSDPVARRVYLTTQLKQKKSTLASVNIQALPDKGQKLIKQIQELEEVLSGLTLSPEQGTNEKSNSQVPQQSHFTKTTTGPPHLVPPQPLPRRGTQPVGSLELK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8458
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.