missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF-1 / NKX2-1 Mouse anti-Human, Mouse, Rat, Clone: rNX2.1/690, Novus Biologicals™
Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | TTF-1 / NKX2-1 |
|---|---|
| Clone | rNX2.1/690 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
TTF-1 / NKX2-1 Monoclonal specifically detects TTF-1 / NKX2-1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| TTF-1 / NKX2-1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones, Tumor Biomarkers | |
| 10 mM PBS with 0.05% BSA | |
| 7080 | |
| IgG1 κ | |
| Protein A or G purified |
| rNX2.1/690 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
| Recombinant full-length human TTF-1/ NKX2-1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title