missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TTF-1/ NKX2-1 Antibody (CL14523), Novus Biologicals™
Mouse Monoclonal Antibody
£298.00 - £449.00
Specifications
| Antigen | TTF-1 / NKX2-1 |
|---|---|
| Clone | CL14523 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043228
|
Novus Biologicals™
NBP3-26860-100ul |
100 μL |
£449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043361
|
Novus Biologicals™
NBP3-26860-25ul |
25 μL |
£298.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| TTF-1 / NKX2-1 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones, Tumor Biomarkers | |
| PBS, pH 7.2, 40% glycerol | |
| 7080 | |
| IgG2a | |
| Protein A purified |
| CL14523 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
| This antibody was developed against a Synthetic Peptide of P43699. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title