missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116246
This item is not returnable.
View return policy
Description
TTC8 Polyclonal Antibody for Western Blot
This gene encodes a protein that has been directly linked to Bardet-Biedl syndrome. The primary features of this syndrome include retinal dystrophy, obesity, polydactyly, renal abnormalities and learning disabilities. Experimentation in non-human eukaryotes suggests that this gene is expressed in ciliated cells and that it is involved in the formation of cilia. Alternate transcriptional splice variants have been characterized.
Specifications
| TTC8 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8TAM2 | |
| TTC8 | |
| KLH conjugated synthetic peptide between 18-46 amino acids from the N-terminal region of human TTC8. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| TTC8 | |
| 0610012F22Rik; AV001447; Bardet-Biedl syndrome 8 protein; Bardet-Biedl syndrome 8 protein homolog; Bardet-Biedl syndrome type 8; BBS8; RP51; tetratricopeptide repeat domain 8; tetratricopeptide repeat protein 8; TPR repeat protein 8; TTC8 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 123016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction