Learn More
Invitrogen™ TTC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589101
Description
Immunogen sequence: MMADEEEEVK PILQKLQELV DQLYSFRDCY FETHSVEDAG RKQQDVQKEM EKTLQQMEEV VGSVQGKAQV LMLTGKALNV TPDYSPKAEE LLSKAVKLEP ELVEAWNQLG EVYWKKGDVA AAHTCFSGAL THCRNKVSLQ NLSMVLRQLR TDTEDEHSHH VMDSVRQAKL AVQMDVHDGR SWYILGNSYL SLYFSTGQNP KISQQALSAY AQAEKVDRKA SSNPDLHLNR ATLHKYEESY GEALEGFSRA AALDPAWPEP RQREQQLLEF LDRLTSLLES KGKVKTKKLQ SMLGSLRPAH; Positive Samples: A-431, MCF-7, Mouse spleen, Mouse testis, Mouse thymus, Rat brain; Cellular Location: Cytoplasm, Nucleus.
Tetratricopeptide repeat protein 5 (TTC5) is a member of a diverse group of functionally distinct proteins that are characterized by containing one or more tetratricopeptide repeats. Each motif consists of two anti-parallel a-helices such that tandem arrays of TPR motifs generate a right-handed helical structure with an amphipathic channel that may serve to accommodate the complementary region of a target protein. While the exact function of TTC5 remains unclear, it is thought that the TPR motifs serve to mediate protein-protein interactions such as those seen with protein chaperones HSP70 and HSP90 and some proteins involved in cell stress response signaling pathways such as protein phosphatase 5, suggesting that TTC5 may also function via protein-protein interactions mediated by its TPR motifs.
Specifications
| TTC5 | |
| Polyclonal | |
| Unconjugated | |
| TTC5 | |
| Strap; stress-responsive activator of p300; tetratricopeptide repeat domain 5; tetratricopeptide repeat protein 5; TPR repeat protein 5; TTC5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 219022, 91875 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.11 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8N0Z6, Q99LG4 | |
| TTC5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TTC5 (NP_6123852). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.