missing translation for 'onlineSavingsMsg'
Learn More

TTC36 Antibody, R&D Systems™

Product Code. 18602615 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18602615 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18602615 Supplier Novus Biologicals Supplier No. NBP323480
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

TTC36 Polyclonal specifically detects TTC36 in Human samples. It is validated for Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC36
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 0.5-1 ul/mg of lysate
Formulation PBS, pH 7.0
Gene Alias HBP21, tetratricopeptide repeat domain 36, tetratricopeptide repeat protein 36
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TTC36 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143941
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.