missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554617
This item is not returnable.
View return policy
Description
Immunogen sequence: ELYDVTWEEM RDKMRKWREE NSRNSEQIVE VGEELINEYA SKLGDDIWII YEQVMIAALD YGRDDLALFC LQELRRQFPG SH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 73%.
The exact function of TTC35 remains unknown.
Specifications
| TTC35 | |
| Polyclonal | |
| Unconjugated | |
| Emc2 | |
| 4921531G14Rik; AV060620; AW209495; emc2; ER membrane protein complex subunit 2; KIAA0103; KIAA0103-like protein; RGD1310430; tetratricopeptide repeat domain 35; tetratricopeptide repeat protein 35; Tetratricopeptide repeat protein 35-like protein; TPR repeat protein 35; TTC35; Unknown (protein for MGC:128301); zgc:73154; zgc:86891 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9694 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15006 | |
| Emc2 | |
| Recombinant protein corresponding to Human TTC35. Recombinant protein control fragment (Product #RP-93768). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction