missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC33 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£190.00 - £255.00
Specifications
| Antigen | TTC33 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620956
|
Novus Biologicals
NBP2-99640-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651326
|
Novus Biologicals
NBP2-99640-50ul |
50 μg |
£190.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTC33 Polyclonal antibody specifically detects TTC33 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TTC33 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.0) | |
| 23548 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| osmosis responsive factor, Osmosis-responsive factor, OSRF, tetratricopeptide repeat domain 33, tetratricopeptide repeat protein 33, TPR repeat protein 33 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TTC33. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title