missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TTC33 Polyclonal antibody specifically detects TTC33 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Purification Method | Antigen and protein A Affinity-purified |
| Regulatory Status | RUO |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?