missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC33 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97329APC
This item is not returnable.
View return policy
Description
TTC33 Polyclonal antibody specifically detects TTC33 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| TTC33 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| osmosis responsive factor, Osmosis-responsive factor, OSRF, tetratricopeptide repeat domain 33, tetratricopeptide repeat protein 33, TPR repeat protein 33 | |
| Produced in rabbits immunized with E. coli-derived Human TTC33 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 23548 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction