missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557912
This item is not returnable.
View return policy
Description
Immunogen sequence: IDCGKKEAEA HMHMGLLYEE DGQLLEAAEH YEAFHQLTQG RIWKDETGRS LNLLACESLL RTYRLLSDKM LENKEYKQAI KILIKA Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
TTC29 is a protein coding gene.
Specifications
| TTC29 | |
| Polyclonal | |
| Unconjugated | |
| TTC29 | |
| 1700031F13Rik; NYD-SP14; Protein TBPP2A; TBPP2A; testicular secretory protein Li 62; testis development protein NYD-SP14; tetratricopeptide repeat domain 29; tetratricopeptide repeat protein 29; TPR repeat protein 29; TTC29 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83894 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NA56 | |
| TTC29 | |
| Recombinant protein corresponding to Human TTC29. Recombinant protein control fragment (Product #RP-96653). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction