Learn More
TTC28 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25701-1-AP-150UL
This item is not returnable.
View return policy
Description
The TTC28 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TTC28. This antibody recognizes human antigen. The TTC28 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| TTC28 | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunoprecipitation, ELISA | |
| KIAA1043, TPR repeat protein 28, TTC28 | |
| 150 μL | |
| 23331 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |