missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556262
This item is not returnable.
View return policy
Description
Immunogen sequence: QVLEKSRRLA ERSTGRRLLG HLYNDLGLGY SQLQLFPLAV EAFLQALPLC WVPGEQATVL RNLGMAHNAL GNYQEAREFH QKAADLHGSV G Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 72%.
Specifications
| TTC24 | |
| Polyclonal | |
| Unconjugated | |
| TTC24 | |
| tetratricopeptide repeat domain 24; tetratricopeptide repeat protein 24; TPR repeat protein 24; TTC24 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 164118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A2A3L6 | |
| TTC24 | |
| Recombinant protein corresponding to Human TTC24. Recombinant protein control fragment (Product #RP-94527). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction