missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568867
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%; Rat: 79%; Yeast: 92%.
This gene encodes a protein of unknown function. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.
Specifications
| TTC19 | |
| Polyclonal | |
| Unconjugated | |
| TTC19 | |
| 2010204O13Rik; 2810460C24Rik; AI505442; MC3DN2; RGD1311797; tetratricopeptide repeat domain 19; tetratricopeptide repeat protein 19, mitochondrial; tetratricopeptide repeat protein 19; tetratricopeptide repeat protein 19, mitochondrial; TPR repeat protein 19; TTC19 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 72795 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8CC21 | |
| TTC19 | |
| synthetic peptide directed towards the N terminal region of mouse TTC19. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction