missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC19 Polyclonal Antibody

TTC19 Polyclonal Antibody
Brand: Thermo Scientific PA5112295
This item is not returnable.
View return policy
Description
TTC19 Polyclonal Antibody for Western Blot, ICC/IF, IP
Specifications
| TTC19 | |
| Polyclonal | |
| Unconjugated | |
| TTC19 | |
| 2010204O13Rik; 2810460C24Rik; AI505442; MC3DN2; RGD1311797; tetratricopeptide repeat domain 19; tetratricopeptide repeat protein 19, mitochondrial; tetratricopeptide repeat protein 19; tetratricopeptide repeat protein 19, mitochondrial; TPR repeat protein 19; TTC19 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 54902 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q6DKK2 | |
| TTC19 | |
| E. coli-derived Human TTC19 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction