missing translation for 'onlineSavingsMsg'
Learn More

TTC15 Antibody, Novus Biologicals™

Product Code. 18327079 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327079 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327079 Supplier Novus Biologicals Supplier No. H00051112B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TTC15 Polyclonal antibody specifically detects TTC15 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC15
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence 10 ug/mL
Formulation PBS (pH 7.4)
Gene Accession No. AAH14164.2
Gene Alias CGI-87, FLJ36862, FLJ44560, tetratricopeptide repeat domain 15, tetratricopeptide repeat protein 15, TPR repeat protein 15
Host Species Mouse
Immunogen TTC15 (AAH14164.2, 1 a.a. - 735 a.a.) full-length human protein. MEDAGGGEETPAPEAPHPPQLAPPEEQGLLFQEETIDLGGDEFGSEENETASEGSSPLADKLNEHMMESVLISDSPNSEGDAGDLGRVRDEAEPGGEGDPGPEPAGTPSPSGEADGDCAPEDAAPSSGGAPRQDAAREVPGSEAARPEQEPPVAEPVPVCTIFSQRAPPASGDGFEPQMVKSPSFGGASEASARTPPQVVQPSPSLSTFFGDTAASHSLASDFFDSFTTSAFISVSNPGAGSPAPASPPPLAVPGTEGRPEPVAMRGPQAAAPPASPEPFAHIQAVFAGSDDPFATALSMSEMDRRNDAWLPGEATRGVLRAVATQQRGAVFVDKENLTMPGLRFDNIQGDAVKDLMLRFLGEKAAAKRQVLNADSVEQSFVGLKQLISCRNWRAAVDLCGRLLTAHGQGYGKSGLLTSHTTDSLQLWFVRLALLVKLGLFQNAEMEFEPFGNLDQPDLYYEYYPHVYPGRRGSMVPFSMRILHAELQQYLGNPQESLDRLHKVKTVCSKILANLEQGLAEDGGMSSVTQEGRQASIRLWRSRLGRVMYSMANCLLLMKDYVLAVDAYHSVIKYYPEQEPQLLSGIGRISLQIGDIKTAEKYFQDVEKVTQKLDGLQGKIMVLMNSAFLHLGQNNFAEAHRFFTEILRMDPRNAVANNNAAVCLLYLGKLKDSLRQLEAMVQQDPRHYLHESVLFNLTTMYELESSRSMQKKQALLEAVAGKEGDSFNTQCLKLA
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51112
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.