Learn More
Invitrogen™ TTC14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113721
Description
Immunogen sequence: LLSLLRSEQQ DNPHFRSLLG SAAEPARGPP PQHPLQGRKE KRVDNIEIQK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Rabbit: 79%; Rat: 80%.
TTC14 is a protein coding gene.
Specifications
| TTC14 | |
| Polyclonal | |
| Unconjugated | |
| TTC14 | |
| 2700016E08Rik; 4930434D01Rik; 4931403I22Rik; 4933402I15Rik; AI662468; AU014779; AW561908; cI-44; DRDL5813; Kiaa1980; mKIAA1980; PRO19630; tetratricopeptide repeat domain 14; tetratricopeptide repeat protein 14; TPR repeat protein 14; Ttc14; UNQ5813; UNQ5813/PRO19630 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 151613 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96N46 | |
| TTC14 | |
| Synthetic peptide directed towards the N terminal region of human TTC14 (aa 17-66). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.