missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC14 Antibody (OTI9E11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74655F
This item is not returnable.
View return policy
Description
TTC14 Monoclonal specifically detects TTC14 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| TTC14 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TTC14 | |
| Recombinant protein fragment corresponding to amino acids 429-770 of human TTC14 (NP_597719) produced in E.coli. | |
| 0.1 mL | |
| 151613.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI9E11 | |
| Immunohistochemistry | |
| DKFZp313M1015, DRDL5813, KIAA1980FLJ00166, PRO19630, tetratricopeptide repeat domain 14, tetratricopeptide repeat protein 14, TPR repeat protein 14 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction