missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544109
This item is not returnable.
View return policy
Description
Peptide sequence: MNLCLQAPFV SEVWAVEVSR RCLSLLNSQD GGILTRAAGV LSRTLSSSLK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
TTC12 contains 3 TPR repeats. Hypermethylation of TTC12 gene may play a role in acute lymphoblastic leukemia. Haplotypic variants in DRD2, ANKK1, TTC12, and NCAM1 are associated with comorbid alcohol and drug dependence.
Specifications
| TTC12 | |
| Polyclonal | |
| Unconjugated | |
| TTC12 | |
| E330017O07Rik; tetratricopeptide repeat domain 12; tetratricopeptide repeat protein 12; TPARM; TPR repeat protein 12; TTC12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54970 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H892 | |
| TTC12 | |
| Synthetic peptide directed towards the C-terminal of human TTC12 (aa 504-553). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction